Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500018260.mm.1 |
TC0500002008.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018260.mm.1 |
NM_001080977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018260.mm.1 |
NM_001080977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018262.mm.1 |
TC0500002008.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018262.mm.1 |
NM_001080977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018262.mm.1 |
NM_001080977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018256.mm.1 |
TC0500002008.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018256.mm.1 |
NM_001080977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018256.mm.1 |
NM_001080977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018257.mm.1 |
TC0500002008.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018257.mm.1 |
NM_001080977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018257.mm.1 |
NM_001080977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018258.mm.1 |
TC0500002008.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018258.mm.1 |
NM_001080977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018258.mm.1 |
NM_001080977.1 |
Refseq |