Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133758.3 | NP_598519.2 | 16 - 17 | 2062 | 98 | |
NM_177249.3 | NP_796223.2 | 17 - 18 | 2122 | 98 | ||
GenBank mRNA | AK131138.1 | - | 7 - 8 | 739 | 98 | |
BC108425.1 | - | 17 - 18 | 2063 | 30062566 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |