Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178347.4 | NP_848124.1 | 13 - 14 | 1457 | 67 | |
GenBank mRNA | AK038523.1 | - | 13 - 14 | 1440 | 67 | |
AK047789.1 | - | 13 - 14 | 1440 | 67 | ||
AK048246.1 | - | 13 - 14 | 1440 | 67 | ||
AK052567.1 | - | 13 - 14 | 1457 | 67 | ||
AK075948.1 | - | 13 - 14 | 1458 | 67 | ||
AK081461.1 | - | 13 - 14 | 1458 | 67 | ||
AK169677.1 | - | 9 - 10 | 931 | 67 | ||
AK219876.1 | - | 1 - 2 | 37 | 67 | ||
BC059013.1 | - | 13 - 14 | 1426 | 6813730 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |