Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081267.2 | NP_001074736.1 | 8 - 9 | 93 | ||
GenBank mRNA | AK032853.1 | - | 8 - 9 | 93 | ||
AK146675.1 | - | 8 - 9 | 93 | |||
BC054454.1 | - | 3 - 4 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |