Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028145.1 | NP_082421.1 | 3 - 4 | 1143 | 61 | |
GenBank mRNA | AK013012.1 | - | 3 - 4 | 1121 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Mus musculus Probe spans exons |