Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026165.3 | NP_080441.1 | 1 - 2 | 369 | 95 | |
GenBank mRNA | AK004625.1 | - | 1 - 2 | 373 | 95 | |
AK008131.1 | - | 1 - 2 | 373 | 95 | ||
AK030346.1 | - | 1 - 2 | 370 | 95 | ||
AK031117.1 | - | 1 - 2 | 370 | 95 | ||
AK078383.1 | - | 379 | 95 | |||
AK147936.1 | - | 1 - 2 | 384 | 95 | ||
AK160834.1 | - | 1 - 2 | 370 | 95 | ||
BC020087.1 | - | 1 - 2 | 353 | 3990909 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |