Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019442.3 | NP_062315.1 | 1 - 2 | 127 | 142 | |
GenBank mRNA | AF099934.1 | - | 1 - 2 | 61 | 142 | |
AK146739.1 | - | 1 - 2 | 129 | 142 | ||
AK168865.1 | - | 1 - 2 | 129 | 142 | ||
BC022681.1 | - | 1 - 2 | 127 | 4240347 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |