Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181588.3 | NP_853619.1 | 4 - 5 | 514 | 121 | |
GenBank mRNA | BC024580.1 | - | 4 - 5 | 538 | 5064316 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |