Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170638.1 | NP_001164109.1 | 11 - 12 | 1384 | 96 | |
NM_009198.3 | NP_033224.3 | 11 - 12 | 1476 | 96 | ||
NR_121611.1 | 11 - 12 | 1955 | 96 | |||
GenBank mRNA | AK132800.1 | - | 11 - 12 | 1955 | 96 | |
AK164712.1 | - | 11 - 12 | 1386 | 96 | ||
BC013445.1 | - | 11 - 12 | 1417 | 4241557 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |