Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111110.2 | NP_001104580.1 | 8 - 9 | 1224 | 77 | |
NM_001284519.1 | NP_001271448.1 | 9 - 10 | 1386 | 77 | ||
NM_001284520.1 | NP_001271449.1 | 7 - 8 | 879 | 77 | ||
NM_007717.5 | NP_031743.3 | 7 - 8 | 879 | 77 | ||
GenBank mRNA | AB061276.1 | - | 8 - 9 | 1208 | 77 | |
AB061277.1 | - | 9 - 10 | 1370 | 77 | ||
AK083394.1 | - | 5 - 6 | 722 | 77 | ||
AK145110.1 | - | 6 - 7 | 1027 | 77 | ||
AK166486.1 | - | 7 - 8 | 880 | 77 | ||
BC028474.1 | - | 8 - 9 | 1234 | 77 | ||
BC055079.1 | - | 6 - 7 | 876 | 30077348 | 77 | |
D21826.1 | - | 6 - 7 | 851 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |