Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134069.3 | NP_598830.2 | 2 - 3 | 356 | 98 | |
GenBank mRNA | AK128896.1 | - | 2 - 3 | 340 | 98 | |
AK143896.1 | - | 2 - 3 | 328 | 98 | ||
AK165505.1 | - | 2 - 3 | 201 | 98 | ||
BC018252.1 | - | 2 - 3 | 285 | 4240254 | 98 | |
BC024459.1 | - | 2 - 3 | 312 | 4975341 | 98 | |
BC034179.1 | - | 2 - 3 | 176 | 4972500 | 98 | |
BC034183.1 | - | 2 - 3 | 161 | 4973620 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |