Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013491.2 | NP_038519.1 | 2 - 3 | 382 | 106 | |
GenBank mRNA | AK036735.1 | - | 2 - 3 | 384 | 106 | |
AY046403.1 | - | 2 - 3 | 207 | 106 | ||
AY046404.1 | - | 2 - 3 | 207 | 106 | ||
BC114336.1 | - | 2 - 3 | 347 | 40048966 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |