Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011188.3 | NP_035318.1 | 6 - 7 | 826 | 77 | |
GenBank mRNA | AK005083.1 | - | 5 - 6 | 449 | 77 | |
AK076874.1 | - | 6 - 7 | 828 | 77 | ||
AK146919.1 | - | 5 - 6 | 467 | 77 | ||
AK153413.1 | - | 5 - 6 | 470 | 77 | ||
AK168548.1 | - | 5 - 6 | 467 | 77 | ||
AK216249.1 | - | 2 - 3 | 77 | |||
BC005462.1 | - | 5 - 6 | 476 | 3590739 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |