Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hipk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001294143.1 | NP_001281072.1 | 1 - 2 | 399 | 111 | |
NM_001294144.1 | NP_001281073.1 | 1 - 2 | 399 | 111 | ||
NM_010433.2 | NP_034563.2 | 1 - 2 | 399 | 111 | ||
GenBank mRNA | AF077659.1 | - | 1 - 2 | 162 | 111 | |
AF170301.1 | - | 1 - 2 | 149 | 111 | ||
AF170302.1 | - | 1 - 2 | 149 | 111 | ||
AF273680.1 | - | 1 - 2 | 120 | 111 | ||
BC031904.1 | - | 1 - 2 | 132 | 5368577 | 111 | |
CJ173672.1 | - | 1 - 2 | 399 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |