Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023626.4 | NP_076115.3 | 4 - 5 | 419 | 72 | |
GenBank mRNA | AK030364.1 | - | 3 - 4 | 226 | 72 | |
AK080787.1 | - | 4 - 5 | 388 | 72 | ||
AK131993.1 | - | 4 - 5 | 419 | 72 | ||
AK146145.1 | - | 3 - 4 | 236 | 72 | ||
AY007791.1 | - | 4 - 5 | 388 | 72 | ||
BC018342.1 | - | 4 - 5 | 369 | 4010253 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |