Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172120.4 | NP_742118.3 | 23 - 24 | 1997 | 94 | |
GenBank mRNA | AB028843.1 | - | 23 - 24 | 1997 | 94 | |
AF327407.1 | - | 23 - 24 | 1973 | 94 | ||
AK051658.1 | - | 23 - 24 | 1992 | 94 | ||
AK162208.1 | - | 23 - 24 | 1996 | 94 | ||
AK164177.1 | - | 23 - 24 | 1991 | 94 | ||
BC005530.1 | - | 21 - 22 | 1864 | 94 | ||
BC023243.1 | - | 22 - 23 | 1963 | 94 | ||
BC049916.1 | - | 22 - 23 | 1970 | 94 | ||
BC119362.1 | - | 23 - 24 | 1992 | 8734111 | 94 | |
BC125299.1 | - | 23 - 24 | 1992 | 40129814 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |