Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rala | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019491.5 | NP_062364.3 | 2 - 3 | 346 | 80 | |
GenBank mRNA | AF244951.1 | - | 1 - 2 | 109 | 80 | |
AK013881.1 | - | 2 - 3 | 335 | 80 | ||
BC031741.1 | - | 2 - 3 | 346 | 4501311 | 80 | |
Z48587.1 | - | 1 - 2 | 109 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |