Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019429.2 | NP_062302.1 | 5 - 6 | 632 | 78 | |
GenBank mRNA | AJ131775.1 | - | 5 - 6 | 632 | 78 | |
AK088019.1 | - | 5 - 6 | 660 | 78 | ||
BC125481.1 | - | 5 - 6 | 630 | 40129996 | 78 | |
BC131955.1 | - | 5 - 6 | 630 | 40130232 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |