Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008156.2 | NP_032182.2 | 7 - 8 | 805 | 92 | |
GenBank mRNA | AF050666.1 | - | 7 - 8 | 747 | 92 | |
AK004957.2 | - | 6 - 7 | 509 | 92 | ||
AY081194.1 | - | 6 - 7 | 532 | 92 | ||
AY333960.1 | - | 6 - 7 | 489 | 92 | ||
BC019146.1 | - | 6 - 7 | 518 | 5052822 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |