Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201352.2 | NP_958740.2 | 2 - 3 | 746 | 61 | |
GenBank mRNA | AK141189.1 | - | 2 - 3 | 779 | 61 | |
AK154771.1 | - | 2 - 3 | 883 | 61 | ||
BC024955.1 | - | 1 - 2 | 32 | 4953174 | 61 | |
BC082585.1 | - | 2 - 3 | 746 | 30608279 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |