Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011035.2 | NP_035165.2 | 3 - 4 | 519 | 61 | |
GenBank mRNA | AF082077.1 | - | 3 - 4 | 481 | 61 | |
AK077497.1 | - | 3 - 4 | 485 | 61 | ||
BC055740.1 | - | 3 - 4 | 523 | 61 | ||
BC060157.1 | - | 3 - 4 | 523 | 61 | ||
BC079838.1 | - | 3 - 4 | 505 | 61 | ||
BC145698.1 | - | 4 - 5 | 443 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |