Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfibp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163557.1 | NP_001157029.1 | 3 - 4 | 407 | 94 | |
NM_008905.2 | NP_032931.2 | 3 - 4 | 643 | 94 | ||
GenBank mRNA | AK035364.1 | - | 3 - 4 | 411 | 94 | |
AK135964.1 | - | 3 - 4 | 678 | 94 | ||
BB627669.1 | - | 3 - 4 | 408 | 94 | ||
BC051202.1 | - | 3 - 4 | 491 | 94 | ||
BC086931.1 | - | 3 - 4 | 762 | 94 | ||
BC100346.1 | - | 3 - 4 | 407 | 30936010 | 94 | |
U79024.1 | - | 3 - 4 | 428 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |