Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wasl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028459.2 | NP_082735.2 | 5 - 6 | 806 | 133 | |
GenBank mRNA | AJ318416.1 | - | 5 - 6 | 524 | 133 | |
AK140189.1 | - | 5 - 6 | 724 | 133 | ||
AK142642.1 | - | 4 - 5 | 282 | 133 | ||
AK143969.1 | - | 5 - 6 | 787 | 133 | ||
AK144789.1 | - | 5 - 6 | 782 | 133 | ||
AK147881.1 | - | 3 - 4 | 193 | 133 | ||
AK159052.1 | - | 5 - 6 | 758 | 133 | ||
BC019951.1 | - | 5 - 6 | 696 | 133 | ||
BC055045.1 | - | 5 - 6 | 782 | 6489610 | 133 | |
BC058642.1 | - | 5 - 6 | 806 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |