Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wasl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028459.2 | NP_082735.2 | 8 - 9 | 1164 | 121 | |
GenBank mRNA | AJ318416.1 | - | 8 - 9 | 882 | 121 | |
AK142642.1 | - | 7 - 8 | 640 | 121 | ||
AK143969.1 | - | 8 - 9 | 1145 | 121 | ||
AK147881.1 | - | 6 - 7 | 551 | 121 | ||
AK159052.1 | - | 8 - 9 | 1116 | 121 | ||
BC055045.1 | - | 8 - 9 | 1140 | 6489610 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |