Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025350.3 | NP_079626.2 | 3 - 4 | 622 | 70 | |
GenBank mRNA | AK003088.1 | - | 3 - 4 | 407 | 70 | |
BC052661.1 | - | 3 - 4 | 636 | 30050094 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |