Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nucb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130479.2 | NP_001123951.1 | 7 - 8 | 957 | 72 | |
GenBank mRNA | AJ222586.1 | - | 7 - 8 | 957 | 72 | |
AK145934.1 | - | 7 - 8 | 912 | 72 | ||
BC010459.1 | - | 7 - 8 | 871 | 3585076 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |