Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nucb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130479.2 | NP_001123951.1 | 9 - 10 | 1108 | 86 | |
GenBank mRNA | AJ222586.1 | - | 9 - 10 | 1108 | 86 | |
AK145934.1 | - | 9 - 10 | 1063 | 86 | ||
AK152672.1 | - | 3 - 4 | 345 | 86 | ||
BC010459.1 | - | 9 - 10 | 1022 | 3585076 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |