Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170744.1 | NP_001164215.1 | 6 - 7 | 2585 | 67 | |
NM_009980.4 | NP_034110.1 | 8 - 9 | 1462 | 67 | ||
GenBank mRNA | AB033123.1 | - | 8 - 9 | 1327 | 67 | |
AF016508.1 | - | 6 - 7 | 934 | 67 | ||
AK134399.1 | - | 7 - 8 | 986 | 67 | ||
AK146713.1 | - | 8 - 9 | 1347 | 67 | ||
AK147871.1 | - | 8 - 9 | 1345 | 67 | ||
BC013333.1 | - | 6 - 7 | 969 | 4019572 | 67 | |
BC017520.1 | - | 7 - 8 | 2430 | 4511826 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |