Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011212.3 | NP_035342.3 | 12 - 13 | 1330 | 68 | |
GenBank mRNA | AK004602.1 | - | 12 - 13 | 1332 | 68 | |
AK132455.1 | - | 11 - 12 | 1293 | 68 | ||
AK154910.1 | - | 9 - 10 | 882 | 68 | ||
AK160947.1 | - | 9 - 10 | 879 | 68 | ||
BC053408.1 | - | 12 - 13 | 1106 | 68 | ||
BC098473.1 | - | 12 - 13 | 1094 | 68 | ||
D83484.1 | - | 12 - 13 | 1667 | 68 | ||
U35368.1 | - | 12 - 13 | 1330 | 68 | ||
U36758.1 | - | 9 - 10 | 879 | 68 | ||
U40280.1 | - | 11 - 12 | 1070 | 68 | ||
U62387.1 | - | 12 - 13 | 1232 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |