Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Athl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145387.4 | NP_663362.2 | 10 - 11 | 1705 | 80 | |
GenBank mRNA | AK077662.1 | - | 10 - 11 | 1706 | 80 | |
AK085600.1 | - | 9 - 10 | 1876 | 80 | ||
BC023151.1 | - | 9 - 10 | 1391 | 80 | ||
BC056953.1 | - | 10 - 11 | 1667 | 5717810 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |