Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143755.1 | NP_001137227.1 | 3 - 4 | 516 | 80 | |
NM_027164.1 | NP_081440.1 | 2 - 3 | 561 | 80 | ||
GenBank mRNA | BC050816.1 | - | 2 - 3 | 561 | 6774506 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |