Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841688 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841686 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841705 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841684 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841610 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841611 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841613 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841687 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841685 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841612 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2002557 |
BC056922.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2002557 |
NM_011480.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2002557 |
AK040663.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2002557 |
AK052628.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
106929 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
610342 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
515333 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
457530 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829309 |
AK031119.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829309 |
AK054259.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829309 |
AK081004.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829309 |
NM_178782.4 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
952056 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914973 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
374916 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
748445 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
811343 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
681482 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
341943 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
59677 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
76199 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
95480 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P141512 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P138617 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2002557 |
BC056922.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2002557 |
NM_011480.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2002557 |
AK040663.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2002557 |
AK052628.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P401921 |
BC006051.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P401921 |
BC056922.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P401921 |
AK158269.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P401921 |
AK169607.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P401921 |
AK150052.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841717 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841732 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841678 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841671 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841690 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841676 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841719 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841721 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841697 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841729 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841683 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841724 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841675 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841681 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841705 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841693 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841673 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841686 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841691 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841728 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841701 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841707 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841713 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841725 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841715 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841709 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841703 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841730 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841708 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841687 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841680 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841679 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841698 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841716 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841685 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841700 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841688 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841677 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27841712 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07361982 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P138617 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P141512 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02338790 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P401921 |
BC006051.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P401921 |
BC056922.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P401921 |
AK158269.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P401921 |
AK169607.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P401921 |
AK150052.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P101311 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P141512 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P138617 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09264451 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09264439 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09264441 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09264452 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09422801 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09264440 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841688 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841684 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841686 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841611 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841705 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841613 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841685 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841687 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841610 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27841612 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841599 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841536 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841614 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841540 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841730 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841658 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841650 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841671 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841624 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841705 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841604 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841690 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841627 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841729 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841542 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841625 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841636 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841557 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841681 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841522 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841572 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841725 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841717 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841527 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841688 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841618 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841646 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841568 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841583 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841530 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841701 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841520 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841622 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841621 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841639 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841566 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841683 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841584 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841603 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841721 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841733 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841697 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841637 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841512 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841619 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841686 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841561 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841676 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841611 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841565 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841673 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841633 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841550 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841694 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841709 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841712 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841615 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841678 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841538 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841609 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841573 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841644 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841680 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841597 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841601 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841515 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841708 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841560 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841581 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841631 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841629 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841571 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841593 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841524 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841713 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841700 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841677 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841656 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841617 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841691 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841728 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841595 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841719 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841546 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841555 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841511 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841716 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841623 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841579 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841518 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841535 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841608 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841591 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841606 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841660 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841558 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841663 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841553 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841647 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841576 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841509 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841707 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841569 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841513 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27841533 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09264443 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841646 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841675 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841717 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09078857 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09078856 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09078858 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841700 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841686 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841644 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09644094 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841709 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841697 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09230787 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841730 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09497890 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841609 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09230789 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841716 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841663 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841681 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841641 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09422805 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841660 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841691 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841604 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841603 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841705 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841721 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09644093 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09231716 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841635 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841639 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841632 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841724 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841712 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09230779 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841676 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841690 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09327190 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841708 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09264451 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841647 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841656 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841701 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841713 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09264445 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841615 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841703 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841707 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841618 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841606 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09231714 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841671 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841650 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841640 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841611 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841673 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09264436 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841667 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09141843 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841733 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09078859 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09644084 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841619 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841679 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841728 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27841680 |
- |
- |