Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242349.1 | NP_001229278.1 | 8 - 9 | 1130 | 107 | |
NM_178642.5 | NP_848757.4 | 8 - 9 | 1127 | 107 | ||
GenBank mRNA | AK032793.1 | - | 3 - 4 | 138 | 107 | |
AK052589.1 | - | 8 - 9 | 1079 | 107 | ||
BC062959.1 | - | 8 - 9 | 1016 | 30547439 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |