Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028477.2 | NP_082753.1 | 3 - 4 | 302 | 88 | |
GenBank mRNA | AF525158.1 | - | 3 - 4 | 259 | 88 | |
AK005485.1 | - | 3 - 4 | 303 | 88 | ||
AK014414.1 | - | 3 - 4 | 303 | 88 | ||
BC109185.1 | - | 3 - 4 | 301 | 40050251 | 88 | |
BC109186.1 | - | 3 - 4 | 301 | 40050253 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |