Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152922.3 | NP_690886.3 | 7 - 8 | 774 | 175 | |
GenBank mRNA | AK148723.1 | - | 7 - 8 | 775 | 85 | |
BC139484.1 | - | 7 - 8 | 743 | 8862506 | 175 | |
BC141312.1 | - | 7 - 8 | 743 | 9055924 | 175 | |
BC145632.1 | - | 7 - 8 | 743 | 9054200 | 175 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |