Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmpcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028431.2 | NP_082707.1 | 5 - 6 | 665 | 94 | |
GenBank mRNA | AK013995.1 | - | 5 - 6 | 666 | 94 | |
AK152673.1 | - | 5 - 6 | 666 | 94 | ||
AK169417.1 | - | 5 - 6 | 622 | 94 | ||
AK191207.1 | - | 1 - 2 | 125 | 94 | ||
BC038321.1 | - | 4 - 5 | 536 | 94 | ||
BC051216.1 | - | 4 - 5 | 472 | 94 | ||
BC060662.1 | - | 5 - 6 | 658 | 94 | ||
BC065095.1 | - | 5 - 6 | 658 | 94 | ||
BC087943.1 | - | 5 - 6 | 677 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |