Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175375.3 | NP_780584.2 | 1 - 2 | 451 | 148 | |
GenBank mRNA | AK131166.1 | - | 1 - 2 | 314 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |