Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171147.1 | NP_001164618.1 | 7 - 8 | 1322 | 71 | |
NM_009534.3 | NP_033560.1 | 6 - 7 | 1274 | 71 | ||
GenBank mRNA | BC014733.1 | - | 7 - 8 | 1319 | 4239820 | 71 |
BC039125.1 | - | 6 - 7 | 1129 | 2649070 | 71 | |
BC094313.1 | - | 6 - 7 | 1262 | 5697882 | 71 | |
X80508.1 | - | 6 - 7 | 1266 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |