Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027889.1 | NP_082165.1 | 4 - 5 | 669 | 71 | |
GenBank mRNA | AK004695.1 | - | 4 - 5 | 644 | 71 | |
AK032484.1 | - | 4 - 5 | 634 | 71 | ||
AK037658.1 | - | 4 - 5 | 643 | 71 | ||
AK047228.1 | - | 4 - 5 | 477 | 71 | ||
AK090030.1 | - | 4 - 5 | 650 | 71 | ||
AK153963.1 | - | 4 - 5 | 642 | 71 | ||
BC029004.1 | - | 4 - 5 | 669 | 5360111 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |