Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291915.1 | NP_001278844.1 | 2 - 3 | 282 | 109 | |
NM_013459.3 | NP_038487.1 | 2 - 3 | 285 | 109 | ||
GenBank mRNA | AK143817.1 | - | 2 - 3 | 229 | 109 | |
BC138778.1 | - | 2 - 3 | 285 | 8861800 | 109 | |
BC138779.1 | - | 2 - 3 | 285 | 8861801 | 109 | |
BC145412.1 | - | 2 - 3 | 282 | 9053970 | 109 | |
DV051476.1 | - | 2 - 3 | 230 | 109 | ||
M11768.1 | - | 2 - 3 | 226 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |