Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079824.1 | NP_001073293.1 | 8 - 9 | 1022 | 66 | |
GenBank mRNA | BC147496.1 | - | 6 - 7 | 679 | 66 | |
BX514270.1 | - | 3 - 4 | 251 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |