Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175450.4 | NP_780659.2 | 8 - 9 | 1113 | 96 | |
GenBank mRNA | AK050106.1 | - | 8 - 9 | 1111 | 96 | |
AK082695.1 | - | 8 - 9 | 1116 | 96 | ||
AK133716.1 | - | 8 - 9 | 1114 | 96 | ||
AK160713.1 | - | 8 - 9 | 1111 | 96 | ||
AK166113.1 | - | 1112 | 96 | |||
AK168691.1 | - | 8 - 9 | 1111 | 96 | ||
BC032968.1 | - | 7 - 8 | 805 | 96 | ||
BC095984.1 | - | 8 - 9 | 1113 | 4978914 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |