Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reep6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204931.1 | NP_001191860.1 | 3 - 4 | 434 | 65 | |
NM_139292.2 | NP_647453.1 | 3 - 4 | 434 | 65 | ||
GenBank mRNA | AB039933.1 | - | 3 - 4 | 409 | 65 | |
AK002562.1 | - | 3 - 4 | 432 | 65 | ||
AY562234.1 | - | 3 - 4 | 348 | 65 | ||
BC029741.1 | - | 3 - 4 | 421 | 4195889 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |