Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc51a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145932.3 | NP_666044.1 | 7 - 8 | 1143 | 86 | |
GenBank mRNA | AK052725.1 | - | 7 - 8 | 1144 | 86 | |
AK147155.1 | - | 963 | 86 | |||
BC024441.1 | - | 7 - 8 | 951 | 4973857 | 86 | |
BC025912.1 | - | 7 - 8 | 954 | 5064489 | 86 | |
BC031178.1 | - | 7 - 8 | 894 | 4988213 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |