Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142701.1 | NP_001136173.1 | 6 - 7 | 947 | 108 | |
NM_027521.3 | NP_081797.1 | 5 - 6 | 499 | 108 | ||
GenBank mRNA | AK044860.1 | - | 5 - 6 | 461 | 108 | |
AK151423.1 | - | 6 - 7 | 891 | 108 | ||
AK159194.1 | - | 6 - 7 | 948 | 108 | ||
AK159474.1 | - | 6 - 7 | 880 | 108 | ||
AK170262.1 | - | 6 - 7 | 933 | 108 | ||
AK171304.1 | - | 6 - 7 | 933 | 108 | ||
BC053750.1 | - | 5 - 6 | 499 | 6333971 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |