Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027480.3 | NP_081756.4 | 4 - 5 | 517 | 65 | |
GenBank mRNA | AK043638.1 | - | 4 - 5 | 351 | 65 | |
AK173316.1 | - | 4 - 5 | 340 | 65 | ||
BC048236.1 | - | 3 - 4 | 422 | 6313999 | 65 | |
CB524981.1 | - | 1 - 2 | 46 | 65 | ||
CJ177454.1 | - | 4 - 5 | 356 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |