Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027422.2 | NP_081698.2 | 10 - 11 | 983 | 131 | |
GenBank mRNA | AK154672.1 | - | 10 - 11 | 1000 | 131 | |
AK173128.1 | - | 9 - 10 | 1139 | 131 | ||
BC067013.1 | - | 10 - 11 | 983 | 6810181 | 131 | |
BC080757.1 | - | 3 - 4 | 234 | 131 | ||
BC083321.1 | - | 1 - 2 | 104 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |