Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026665.4 | NP_080941.3 | 4 - 5 | 713 | 72 | |
NR_037622.1 | 4 - 5 | 973 | 72 | |||
GenBank mRNA | AK014862.1 | - | 1 - 2 | 346 | 72 | |
AK014873.1 | - | 4 - 5 | 660 | 72 | ||
AK014982.1 | - | 2 - 3 | 171 | 72 | ||
AK016484.1 | - | 4 - 5 | 497 | 72 | ||
AK019414.1 | - | 4 - 5 | 593 | 72 | ||
AK028408.1 | - | 4 - 5 | 562 | 72 | ||
AK028458.1 | - | 4 - 5 | 541 | 72 | ||
AK129053.1 | - | 4 - 5 | 662 | 72 | ||
AK167523.1 | - | 4 - 5 | 598 | 72 | ||
AY225093.1 | - | 4 - 5 | 506 | 72 | ||
AY251192.1 | - | 4 - 5 | 658 | 72 | ||
BC025588.1 | - | 3 - 4 | 493 | 72 | ||
BC030353.1 | - | 4 - 5 | 984 | 72 | ||
BC050785.1 | - | 2 - 3 | 224 | 6703205 | 72 | |
BC056968.1 | - | 4 - 5 | 959 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |