Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022016.3 | NP_071299.3 | 5 - 6 | 710 | 90 | |
GenBank mRNA | AF229929.1 | - | 5 - 6 | 621 | 90 | |
AF266478.1 | - | 5 - 6 | 756 | 90 | ||
AK020862.1 | - | 4 - 5 | 480 | 90 | ||
BC022970.1 | - | 5 - 6 | 698 | 5366008 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |