Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip5k1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146687.2 | NP_001140159.1 | 12 - 13 | 1607 | 78 | |
NM_001293646.1 | NP_001280575.1 | 12 - 13 | 1607 | 78 | ||
NM_001293647.1 | NP_001280576.1 | 12 - 13 | 1620 | 78 | ||
NM_008844.3 | NP_032870.2 | 12 - 13 | 1607 | 78 | ||
GenBank mRNA | AB006916.1 | - | 12 - 13 | 1590 | 78 | |
AK014403.1 | - | 12 - 13 | 1573 | 78 | ||
AK041402.1 | - | 12 - 13 | 1593 | 78 | ||
AK122319.2 | - | 12 - 13 | 1565 | 78 | ||
AK154816.1 | - | 12 - 13 | 1599 | 78 | ||
AK155371.1 | - | 12 - 13 | 1583 | 78 | ||
AK170286.1 | - | 12 - 13 | 1599 | 78 | ||
AK171576.1 | - | 12 - 13 | 1581 | 78 | ||
BC019138.1 | - | 12 - 13 | 1556 | 5041431 | 78 | |
BC020365.1 | - | 12 - 13 | 1569 | 78 | ||
BC094665.1 | - | 12 - 13 | 1579 | 30356181 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |